Order Entry
United Kingdom
ContactUsLinkComponent
HS-276 is an orally active, potent and highly selective TAK1 inhibitor, with a Ki of 2.5 nM. HS-276 shows significant inhibition of TAK1, CLK2, GCK, ULK2, MAP4K5, IRAK1, NUAK, CSNK1G2, CAMKKβ-1, and MLK1, with IC50 values of 8.25, 29, 33, 63, 125, 26 1 * 25 mg
Catalog # MDEXHY-147141
Supplier:  MEDCHEMEXPRESS
HS-276 is an orally active, potent and highly selective TAK1 inhibitor, with a Ki of 2.5 nM. HS-276 shows significant inhibition of TAK1, CLK2, GCK, ULK2, MAP4K5, IRAK1, NUAK, CSNK1G2, CAMKKβ-1, and MLK1, with IC50 values of 8.25, 29, 33, 63, 125, 26 1 * 25 mg
Catalog # MDEXHY-147141
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together