Order Entry
United Kingdom
ContactUsLinkComponent
SPECIFICATION INCLUDES THE BELOW: CHEMFAST SHARP 18 F00080700000 4 EXTERNAL SOCKETS FX0080721600 INVERTER KIT FOR REMOTE EXTRACTOR – this is needed if the unit is connected to a dedicated exhaust motor FX0080720900 VOLUME CONTROL DAMPER D.250 - this 1 * 1 items
Catalog # FASTFAS/1071/25
Supplier:  FASTER
New Product
SPECIFICATION INCLUDES THE BELOW: CHEMFAST SHARP 18 F00080700000 4 EXTERNAL SOCKETS FX0080721600 INVERTER KIT FOR REMOTE EXTRACTOR – this is needed if the unit is connected to a dedicated exhaust motor FX0080720900 VOLUME CONTROL DAMPER D.250 - this 1 * 1 items
Catalog # FASTFAS/1071/25
Supplier:  FASTER