Order Entry
United Kingdom
ContactUsLinkComponent
SafeFAST Premium 218 Touch with Fixed Stand INCLUDES THE BELOW SPECIFICATIONS AND QUANTITIES: SAFEFAST PREMIUM 218 TOUCH F0A025800000 5 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 5 LED LIGHTING FX0021855600 5 LH SIDE GLASS CAMLOCK 1 ROXTEC 1 * 1 items
  FASTFAS/1304/R3-FS
 :  FASTER
New Product
SafeFAST Premium 218 Touch with Fixed Stand INCLUDES THE BELOW SPECIFICATIONS AND QUANTITIES: SAFEFAST PREMIUM 218 TOUCH F0A025800000 5 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 5 LED LIGHTING FX0021855600 5 LH SIDE GLASS CAMLOCK 1 ROXTEC 1 * 1 items
  FASTFAS/1304/R3-FS
 :  FASTER