3-(2-Chlorophenoxy)propionic acid
Bottle
Catalog # APOSOR470101-100MG
New Product
Supplier: Apollo Scientific
3-(2-Chlorophenoxy)propionic acid
Catalog # APOSOR470101-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 7170-45-8
Jump to:
Product Details & Documents
Synonyms: 3-(2-Chlorophenoxy)propanoic acid
- CAS Number:7170-45-8
- Formula:C₉H₉ClO₃
- MDL Number:MFCD02295728
- Molecular Weight:200.6190 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



