Mal-PEG4-t-butyl ester
Bottle
Catalog # APOSBIPG1517-1G
New Product
Supplier: Apollo Scientific
Mal-PEG4-t-butyl ester
Catalog # APOSBIPG1517-1G
Supplier: Apollo Scientific
New Product
CAS Number: 518044-36-5
Jump to:
Product Details & Documents
Synonyms: tert-Butyl 1-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3,6,9,12-tetraoxapentadecan-15-oate, Mal-PEG4-Boc
- CAS Number:518044-36-5
- Formula:C₁₉H₃₁NO₈
- MDL Number:MFCD22683300
- Molecular Weight:401.451 g/mol
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



