Order Entry
United Kingdom
ContactUsLinkComponent
1 Nr. wall bench assembly 1296mm x 750mm deep x 930mm high, open end to both ends, comprising of the following: • 16mm thick Trespa Toplab Base worktop c-w 50mm high planted upstand as required supported on our standard base unit as detailed below. T 1 * 1 items
  LBSS13547-R1
New Product
1 Nr. wall bench assembly 1296mm x 750mm deep x 930mm high, open end to both ends, comprising of the following: • 16mm thick Trespa Toplab Base worktop c-w 50mm high planted upstand as required supported on our standard base unit as detailed below. T 1 * 1 items
  LBSS13547-R1