Order Entry
United Kingdom
ContactUsLinkComponent
RS PRO RS14 Handheld Digital Multimeter, 10A ac Max, 10A dc Max, 600V ac Max 1 * 1 items
Catalog # RSCO123-1938
Supplier:  RS COMPONENTS
RS PRO RS14 Handheld Digital Multimeter, 10A ac Max, 10A dc Max, 600V ac Max 1 * 1 items
Catalog # RSCO123-1938
Supplier:  RS COMPONENTS
Frequently Bought Together