Order Entry
United Kingdom
ContactUsLinkComponent
SPECIFICATION INCLUDES ALL THE BELOW ITEMS (QUANTITY 1 UNLESS OTHERWISE INDICATED) CFU 4020 Walkin Unit Estimated overall dimensions - 4000x2700x3000mm (WxDxH) Estimated useful dimensions - 4000x2000x2300mm (WxDxH) COOLING COIL LED LIGHTING G3+F9 P 1 * 1 items
Catalog # FASTFAS/1040-CFU
Supplier:  FASTER
New Product
SPECIFICATION INCLUDES ALL THE BELOW ITEMS (QUANTITY 1 UNLESS OTHERWISE INDICATED) CFU 4020 Walkin Unit Estimated overall dimensions - 4000x2700x3000mm (WxDxH) Estimated useful dimensions - 4000x2000x2300mm (WxDxH) COOLING COIL LED LIGHTING G3+F9 P 1 * 1 items
Catalog # FASTFAS/1040-CFU
Supplier:  FASTER