Order Entry
United Kingdom
ContactUsLinkComponent
CABINET HAS THE FOLLOWING SPEC (QTY 1 UNLESS OTHERWISE INDICATED): SAFEFAST PREMIUM 218 TOUCH F0A025800000 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 LED LIGHTING FX0021855600 LH SIDE GLASS CAMLOCK 1 ROXTEC CF16/19 FS0000001917 RIGHT GLASS 1 * 1 items
Catalog # FASTFAS/1304/25-FS
Supplier:  FASTER
New Product
CABINET HAS THE FOLLOWING SPEC (QTY 1 UNLESS OTHERWISE INDICATED): SAFEFAST PREMIUM 218 TOUCH F0A025800000 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 LED LIGHTING FX0021855600 LH SIDE GLASS CAMLOCK 1 ROXTEC CF16/19 FS0000001917 RIGHT GLASS 1 * 1 items
Catalog # FASTFAS/1304/25-FS
Supplier:  FASTER