Walphos, SL-W002-2 (S)-1-[(S)-1-(Diphenylphosphino)ethyl]-2- [2- (diphenylphosphino)phenyl]ferrocene CAS Reg. No. 1854067-25-6 Pur ity min. 97 %, ee min. 99 % - 1g 1 * 1 g
Catalog # MSPPALG11A03ZZ
Supplier: MARKET SOURCE PART PROCESS
Walphos, SL-W002-2 (S)-1-[(S)-1-(Diphenylphosphino)ethyl]-2- [2- (diphenylphosphino)phenyl]ferrocene CAS Reg. No. 1854067-25-6 Pur ity min. 97 %, ee min. 99 % - 1g 1 * 1 g
Catalog # MSPPALG11A03ZZ
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals