Order Entry
United Kingdom
ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 7x3=21 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 299x42 4x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24016B10
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 7x3=21 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 299x42 4x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24016B10
Supplier:  TENAK