Order Entry
United Kingdom
ContactUsLinkComponent
Inclusive of Items: SafeFAST Premium 218 F0A025800000 1 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 1 LED LIGHTING FX0021855600 1 MAGNETIC UV LAMP FX0002154600 1 LHS GLASS WITH 1 CAMLOCK INLET FX0021248200 1 TOP BOX KIT FOR DECONTAMINATION 1 * 1 items
Catalog # FASTFAS/1309/24
Supplier:  FASTER
New Product
Inclusive of Items: SafeFAST Premium 218 F0A025800000 1 ADDITIONAL EXHAUST INTERNAL HEPA FILTER FX0025841000 1 LED LIGHTING FX0021855600 1 MAGNETIC UV LAMP FX0002154600 1 LHS GLASS WITH 1 CAMLOCK INLET FX0021248200 1 TOP BOX KIT FOR DECONTAMINATION 1 * 1 items
Catalog # FASTFAS/1309/24
Supplier:  FASTER