ZEBRA RIBBON 4800 SERIES WIDTH 4.33 IN LENGTH 242 FT (110MM X 74 M) CORE 0.5 IN THERMAL TRANSFER TECHNOLOGY. COMPATIBLE WITH ZEB RA DESKTOP PRINTERS ZD420, ZD500, ZD620, GC420, GK420, GX420, GX430
Catalog # MSPP366131B
Supplier: MARKET SOURCE PART PROCESS