(R)-PIPERAZINE-2-CARBOXYLIC ACID, 97 1 * 1 g
Catalog # ABCRAB270703-1G
Supplier: abcr
Frequently Bought Together
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
n-Hexane 95%, ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Fisher Scientific
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide, 99.5% for synthesis 1 * 2,5 L
Supplier: PanReac AppliChem
Ethanol absolute 99.8% for analysis
Supplier: Thermo Fisher Scientific
Ethanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
Supplier: Honeywell Chemicals
Glycerine ≥99%
Supplier: Thermo Fisher Scientific
Diethyl ether
Supplier: Roth Carl