Order Entry
United Kingdom
ContactUsLinkComponent
Type: MULTITRON INCUBATOR SHAKER. Triple 3 x variable mm Ø Throw, Top cooling PART CODE INCLUDES: Multitron Triple 3 x variable mm Throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm Power Supply: 230V 50Hz 3 x CO2 Control 3 1 * 1 items
  INFOAN140411
 :  INFORS
New Product
Type: MULTITRON INCUBATOR SHAKER. Triple 3 x variable mm Ø Throw, Top cooling PART CODE INCLUDES: Multitron Triple 3 x variable mm Throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm Power Supply: 230V 50Hz 3 x CO2 Control 3 1 * 1 items
  INFOAN140411
 :  INFORS