Order Entry
United Kingdom
ContactUsLinkComponent
670h x 138w x 450d with 12 apertures of 54mm Rack supplied with 12 storage trays with handgrip Upfold in the base of each tray to locate the box Each rack will hold 60 boxes 51h x 86 x 128mm 1 * 1 items
Catalog # WESBBH629/12NMT
Supplier:  WESBART
New Product
670h x 138w x 450d with 12 apertures of 54mm Rack supplied with 12 storage trays with handgrip Upfold in the base of each tray to locate the box Each rack will hold 60 boxes 51h x 86 x 128mm 1 * 1 items
Catalog # WESBBH629/12NMT
Supplier:  WESBART