Method Pink Grapefruit Hand Wash 354ml 1 * 6 items
Catalog # EVON4004582
Supplier: Deb-STOKO
New Product
Method Pink Grapefruit Hand Wash 354ml 1 * 6 items
Catalog # EVON4004582
Supplier: Deb-STOKO
New Product
Frequently Bought Together
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile with 0,05% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
p-Xylene 99% for spectroscopy
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99% stabilised ethanol-free
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals