Duck Marine Fresh Discs Twin Refill 72ml 1 * 5 items
Catalog # EVON333990
Supplier: Deb-STOKO
New Product
Duck Marine Fresh Discs Twin Refill 72ml 1 * 5 items
Catalog # EVON333990
Supplier: Deb-STOKO
New Product
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Tetramethylammonium chloride, Supelco®
Supplier: Merck
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND