Cutan InstantFOAM Hand Sanitiser - OptiDose 400ml 1 * 12 items
Catalog # EVONCFS400HOP
Supplier: Deb-STOKO
New Product
Cutan InstantFOAM Hand Sanitiser - OptiDose 400ml 1 * 12 items
Catalog # EVONCFS400HOP
Supplier: Deb-STOKO
New Product
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Heptane 99+%, pure
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
Supplier: Honeywell Chemicals
5L Distilled Water 1 * 5 L
Supplier: REAGENT