InstantFOAM Alcohol-Free 47ml 1 * 12 items
Catalog # EVONAFHS47ML
Supplier: Deb-STOKO
New Product
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
1-Butanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Diiodomethane 99+% stabilised
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific