Order Entry
United Kingdom
ContactUsLinkComponent
2 x 25mm Throw 1 x 3mm throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm (20 – 1,000rpm on 3mm) Power Supply: 230V 50Hz 2 x Adhesive mat trays 1 x Microtiter and deep well tray MULTITRON INCUBATOR SHAKER: Triple 2 x 25mm, 1 * 1 items
Catalog # INFOAN139460
Supplier:  INFORS
New Product
2 x 25mm Throw 1 x 3mm throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm (20 – 1,000rpm on 3mm) Power Supply: 230V 50Hz 2 x Adhesive mat trays 1 x Microtiter and deep well tray MULTITRON INCUBATOR SHAKER: Triple 2 x 25mm, 1 * 1 items
Catalog # INFOAN139460
Supplier:  INFORS