(SP-4-1)-[1,3-Bis[2,6-bis(1-methylethyl)phenyl]-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(3-chloropyridine-κN)palladium 1 * 25 g
Catalog # MSPPAG0036US-25G
Supplier: MARKET SOURCE PART PROCESS
(SP-4-1)-[1,3-Bis[2,6-bis(1-methylethyl)phenyl]-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(3-chloropyridine-κN)palladium 1 * 25 g
Catalog # MSPPAG0036US-25G
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals