1-Heptanol, 7-bromo- 1 * 100 g
Catalog # MSPPAG0004XV-100G
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
Supplier: VWR Chemicals
Benzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% for spectrophotometry
Supplier: Thermo Fisher Scientific