Order Entry
United Kingdom
ContactUsLinkComponent
Prototypic, selective kainate receptor agonist. Potent excitant and neurotoxin. 1 * 100 mg
Catalog # MSPPHB0355-100MG
Prototypic, selective kainate receptor agonist. Potent excitant and neurotoxin. 1 * 100 mg
Catalog # MSPPHB0355-100MG
Frequently Bought Together