Order Entry
United Kingdom
ContactUsLinkComponent
SAFE FAST XXL 330 INCLUDING: USEFUL WIDTH: 3300MM USEFUL DEPTH: 1300MM (INCLUDING INFLOW GRIDS) USEFUL HEIGHT: 1300MM WORKING HEIGHT 900MM 8 X POWER SOCKETS (4 ON THE LHS AND 4 ON THE RHS) 2 HOLES ON EACH SIDE WINDOW 19MM DIAMETER FOR CABLES 2 RJ45 S 1 * 1 items
Catalog # FASTFAS/1167/24
Supplier:  FASTER
New Product
SAFE FAST XXL 330 INCLUDING: USEFUL WIDTH: 3300MM USEFUL DEPTH: 1300MM (INCLUDING INFLOW GRIDS) USEFUL HEIGHT: 1300MM WORKING HEIGHT 900MM 8 X POWER SOCKETS (4 ON THE LHS AND 4 ON THE RHS) 2 HOLES ON EACH SIDE WINDOW 19MM DIAMETER FOR CABLES 2 RJ45 S 1 * 1 items
Catalog # FASTFAS/1167/24
Supplier:  FASTER