Microbial DNA Magnetic Extraction employs magnetic particle based purification technology to isolate and purify genomic DNA from bac terial and aspergillus contaminated food plant herbal cannabis matrices and other infused products. Kit Size: 100 Tes 1 * 100 Tests
Catalog # MSPPTBS6025-100
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together