Tris(2-carboxyethyl) phosphine hydrochloride (TCEP) Solution is the broad application to protein and nucleic acid involving the redu ction of disulfide bonds. The pH value of the solution is designed at 6.0, 7.0, and 8.0 for multiple choices. 1 * 10 mL
Catalog # MSPPTBS5082-10-7.0
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together