Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
SoNa™ 520 AM

SoNa™ 520 AM

10 x 50 µg, 50 µG
Catalog # AATB21324
Supplier:   AAT Bioquest
SoNa™ 520 AM
Catalog # AATB21324
Supplier:  AAT Bioquest
CAS Number:  
SoNa™ 520 AM

10 x 50 µg, 50 µG

CAS Number:  
Price
£369.00
Log in for availability
1 * 50 µG
Product Details & Documents

SoNa™ 520 AM is the cell-permeable version of SoNa™ 520 used for monitoring sodium ions in live cells.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:804.8 g/mol
Specifications

Specifications

  • Pk:
    50 µG
  • Pk Info:
    10 x 50 µg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.