Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
SoNa™ 520 AM

SoNa™ 520 AM

1 mg
Catalog # AATB21323
Supplier:   AAT Bioquest
SoNa™ 520 AM
Catalog # AATB21323
Supplier:  AAT Bioquest
CAS Number:  
SoNa™ 520 AM

1 mg

CAS Number:  
Price
£617.00
Log in for availability
1 * 1 mg
Product Details & Documents

SoNa™ 520 AM is the cell-permeable version of SoNa™ 520 used for monitoring sodium ions in live cells.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:804.8 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications