Order Entry
United Kingdom
ContactUsLinkComponent
Multitron Triple 3 x variable mm Throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm Power Supply: 230V 50Hz 3 x CO2 Control 3 x Humidity control and reservoir 3 x Adhesive mat trays 3 x M-tray for Microtitre and deep well p 1 * 1 items
Catalog # INFOAN138273
Supplier:  INFORS
New Product
Multitron Triple 3 x variable mm Throw Temperature Range – 22°C below ambient to 65°C Speed Range: 20 - 400rpm Power Supply: 230V 50Hz 3 x CO2 Control 3 x Humidity control and reservoir 3 x Adhesive mat trays 3 x M-tray for Microtitre and deep well p 1 * 1 items
Catalog # INFOAN138273
Supplier:  INFORS