Order Entry
United Kingdom
ContactUsLinkComponent
Murine RNase Inhibitor 40 U/μl RNR-M5143 is expressed from E. coli cells. It contains AA Met 1 - Ser 456 Accession Q91VI7-1. Functi onal evolution of ribonuclease inhibitor insights from birds and reptiles - PubMed nih.gov 1 * 4 KU
Catalog # ACRBRNR-M5143-4KU
Supplier:  ACROBIOSYSTEMS
Murine RNase Inhibitor 40 U/μl RNR-M5143 is expressed from E. coli cells. It contains AA Met 1 - Ser 456 Accession Q91VI7-1. Functi onal evolution of ribonuclease inhibitor insights from birds and reptiles - PubMed nih.gov 1 * 4 KU
Catalog # ACRBRNR-M5143-4KU
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together