Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Pterostilbene 98%

Glass bottle, 25 g
Catalog # 471380250.
Corrosive
Environment
Pterostilbene 98%
Catalog # 471380250.
CAS Number:  537-42-8
Pterostilbene 98%

Glass bottle, 25 g

CAS Number:  537-42-8
Price
£568.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Formula:C₁₆H₁₆O₃
  • MDL Number:MFCD00238710
  • Molecular Weight:256.2964 g/mol
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    white to beige crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Melting point:
    90°C to 98°C
  • Water:
    ≤3 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.