Order Entry
United Kingdom
ContactUsLinkComponent
of UKAS Reference 7 Points annually. Temperature points -196C,-80C,-20C,+4C,+21C,+37C,+56C, Works undertaken at a UKAS Calibration Lab of a reference handheld thermometer with thermocouple cable @ 7 calibration points, per handheld with single 1 * 1 items
Catalog # TSCAUKAS_T_7
Supplier:  T SCAN
of UKAS Reference 7 Points annually. Temperature points -196C,-80C,-20C,+4C,+21C,+37C,+56C, Works undertaken at a UKAS Calibration Lab of a reference handheld thermometer with thermocouple cable @ 7 calibration points, per handheld with single 1 * 1 items
Catalog # TSCAUKAS_T_7
Supplier:  T SCAN
Frequently Bought Together