Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
C-MYC (HUMAN) RECOMBINANT MONOCLONAL ANTIBODY (9E10) (RAT IGG2CΚ) 1 * 200 µG
Catalog # ENZOENZABS4720200
Supplier:
Enzo Life Sciences
Expand image
C-MYC (HUMAN) RECOMBINANT MONOCLONAL ANTIBODY (9E10) (RAT IGG2CΚ) 1 * 200 µG
Catalog # ENZOENZABS4720200
Supplier:
Enzo Life Sciences
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection