Fluo-8 AM
Plastic bottle, 1.000 µG
Catalog # AATB21083
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
: For research use only.
- CAS Number:1345980-40-6
- Molecular Weight:1046.93 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1.000 µG



