Order Entry
United Kingdom
ContactUsLinkComponent
Bespoke racking: Horizontal outer 615h x 420w x 450d with 11 apertures of 54mm Rack supplied with 33 storage tray with handgrip Upfold in the base of each tray to locate the box Each rack will hold 165 boxes 51h x 86 x 128mm 1 * 1 items
Catalog # WESBBH570/11T
Supplier:  WESBART
Bespoke racking: Horizontal outer 615h x 420w x 450d with 11 apertures of 54mm Rack supplied with 33 storage tray with handgrip Upfold in the base of each tray to locate the box Each rack will hold 165 boxes 51h x 86 x 128mm 1 * 1 items
Catalog # WESBBH570/11T
Supplier:  WESBART
Frequently Bought Together