UN DOSE TYP SDO PE-HD 1600 M 1 * 1 items
SEMA10518
: SEMADENI
Frequently Bought Together
Pipette tips
: Sartorius
VWR®, Beakers, Low Form
: VWR Collection
Buckets with Snap-on Lids
: JOKEY PLASTIK
VWR®, Microtube Storage Boxes
: VWR Collection
Scoops
: VITLAB
Storage Bags
: Alpha Packaging
Storage Boxes
: KOEHLER TECHNISCHE PRODUKTEN
Jars, wide mouth, with screw caps, Nalgene®
: Thermo Fisher Scientific
Organic Premium Cotton Stuff Bag - Natural 1 * 1 items
: MARKET SOURCE PART PROCESS
Robotic tips (CO-RE Tips) and consumables for Microlab
: HAMILTON ROBOTICS
ClipTip™ Racks for Tips
: Thermo Fisher Scientific
Winchester bottles
: PONT PACKAGING
Pressure caps for jars
: LAMAPLAST
Carboys, wide neck, PC, Clearboy™, Nalgene®
: Thermo Fisher Scientific