5,7-dichloro-8-fluoro-2-methylsulfanyl-1H-pyrido[4,3-d]pyrimidin-4-one -100Gram 1 * 100 g
Catalog # PHBLPBX7937-100G
Supplier: Pharmablock (USA)
5,7-dichloro-8-fluoro-2-methylsulfanyl-1H-pyrido[4,3-d]pyrimidin-4-one -100Gram 1 * 100 g
Catalog # PHBLPBX7937-100G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals