NACL 0.05MM SEALED UNIT OMNI 1 * 1 items
Catalog # SPCCGS02221
Supplier: Specac
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Cellulose chromatography papers, Whatman™
Supplier: Whatman products (Cytiva)
384-well high content screening microplates with glass bottom
Supplier: Corning
UNFILTER 96:30 KDA 5-PACK 1 * 5 items
Supplier: MARKET SOURCE PART PROCESS
Bottles, narrow neck, square
Supplier: RIXIUS
Ice pans and buckets, multi-purpose, Corning®
Supplier: Corning
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
PCR plates, 96-well
Supplier: Brand
Accessories and consumables for KingFisher™ Duo Prime
Supplier: Thermo Fisher Scientific
CAP MATS 96 SQUARE WELL 1 * 50 items
Supplier: Thermo Fisher Scientific
BOTTLE PET 350ML RECT STERILE 0.5ML/18MG THIOSULPHATE RED DOME CAP 1 * 174 items
Supplier: LAB3 OWN PRODUCTION
KRYO-BOX CARTON 136X272X50MM 1 * 5 items
Supplier: RATIOLAB
2 x 235 mL deep well plat
Supplier: MARKET SOURCE PART PROCESS
Microtube racks, 96-well
Supplier: HEATHROW
Conway diffusion cell, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP