O1-tert-butyl O2-methyl (2R,4S)-4-aminopiperidine-1,2-dicarboxylate oxalic acid 1 * 1 g
Catalog # PHBLPBZJ8034-3-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O2-methyl (2R,4S)-4-aminopiperidine-1,2-dicarboxylate oxalic acid 1 * 1 g
Catalog # PHBLPBZJ8034-3-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonedimethyl acetal for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
tert-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Chlorobenzene 99.9% for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor