[rel-(3aS,4S,6R,6aR)-4-(hydroxymethyl)-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]methanol 1 * 1 g
Catalog # PHBLPBZ5758-1G
Supplier: Pharmablock (USA)
[rel-(3aS,4S,6R,6aR)-4-(hydroxymethyl)-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]methanol 1 * 1 g
Catalog # PHBLPBZ5758-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane ≥95%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, Technical Grade
Supplier: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals