(2R,4R)-4-fluoro-2-methylpiperidine hydrochloride 1 * 1 g
Catalog # PHBLPBZ4089-1-1G
Supplier: Pharmablock (USA)
(2R,4R)-4-fluoro-2-methylpiperidine hydrochloride 1 * 1 g
Catalog # PHBLPBZ4089-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Heptane anhydrous, 99 % 1 * 1 L
Supplier: Merck
1-Acetyl-2-phenylhydrazine for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Methanol 99.9% for analysis
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Phenetole 98+%
Supplier: Thermo Fisher Scientific
Ethylenediamine, Sigma-Aldrich®
Supplier: Merck