£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
tert-butyl N-[rel-(1S,2S,4S)-4-amino-2-fluoro-cyclohexyl]carbamate hydrochloride 1 * 1 g
PHBLPBZ3783-1-1G
:
Pharmablock (USA)
tert-butyl N-[rel-(1S,2S,4S)-4-amino-2-fluoro-cyclohexyl]carbamate hydrochloride 1 * 1 g
PHBLPBZ3783-1-1G
:
Pharmablock (USA)
Frequently Bought Together
Water, LiChrosolv® tested for UHPLC-MS, Supelco®
: Merck
n-Hexane, EMPLURA®, Supelco®
: Merck
Allyl alcohol for synthesis, Sigma-Aldrich®
: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol absolute for histology, J.T.Baker®
: Avantor
n-Pentane ≥99% (by GC), Riedel-de Haën™
: Honeywell Chemicals
Cyrene™ ≥98.5%, BioRenewable DMF and NMP substitute, Sigma-Aldrich®
: Merck
Acetonitrile with 0,1% ammonium acetate tested for UHPLC-MS, Supelco®
: Merck
Water ACS
: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 40% in aqueous solution, Supelco®
: Merck
CYCLOPENTANE REAGENT GRADE 98% 1 * 500 mL
: Merck
Methanol 99.9% for spectroscopy
: Thermo Fisher Scientific