(2R)-1-[(tert-butoxy)carbonyl]-3,3-difluoropyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBZ2555-1G
Supplier: Pharmablock (USA)
(2R)-1-[(tert-butoxy)carbonyl]-3,3-difluoropyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBZ2555-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Diiodomethane 99+% stabilised
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone, BAKER, J.T.Baker®
Supplier: Avantor
Ferrozine mono-sodium salt hydrate, Supelco®
Supplier: Merck
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% ACS
Supplier: Thermo Fisher Scientific