3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 1 * 10 g
PHBLPBY2010257-10G
Frequently Bought Together
Methanol ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
5L Distilled Water 1 * 5 L
: REAGENT
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
: Honeywell Chemicals
Dichloromethane
: Roth Carl
Toluene, anhydrous ≥99.8%, over molecular sieves
: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Water ACS
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Acetone ≥99%, TECHNICAL
: VWR Chemicals
n-Hexane
: Roth Carl
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
: Thermo Fisher Scientific
Ethylene glycol ≥99%
: Thermo Fisher Scientific