tert-butyl N-[2-[2-[2-[2-[(2-bromoacetyl)amino]ethoxy]ethoxy]ethoxy]ethyl]carbamate 1 * 1 g
PHBLPBY1661-1G
tert-butyl N-[2-[2-[2-[2-[(2-bromoacetyl)amino]ethoxy]ethoxy]ethoxy]ethyl]carbamate 1 * 1 g
PHBLPBY1661-1G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals