(2,3,4,5,6-pentafluorophenyl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate 1 * 1 g
Catalog # PHBLPBY0687-1G
Supplier: Pharmablock (USA)
(2,3,4,5,6-pentafluorophenyl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate 1 * 1 g
Catalog # PHBLPBY0687-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexane
Supplier: Roth Carl
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Amyl Alcohol according to NF V 04-210 for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific