(2S)-6-[[(4S)-5-tert-butoxy-4-(hexadecanoylamino)-5-oxo-pentanoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid 1 * 1 g
Catalog # PHBLPBY0507-1G
Supplier: Pharmablock (USA)
(2S)-6-[[(4S)-5-tert-butoxy-4-(hexadecanoylamino)-5-oxo-pentanoyl]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid 1 * 1 g
Catalog # PHBLPBY0507-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Dibutyl ether anhydrous, 99.3 % 1 * 2 L
Supplier: Merck
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals