[4-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-3-methyl-butanoyl]amino]propanoyl]amino]phenyl]methyl (4-nitrophenyl) carbonate 1 * 1 g
Catalog # PHBLPBY0353-1G
Supplier: Pharmablock (USA)
[4-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-3-methyl-butanoyl]amino]propanoyl]amino]phenyl]methyl (4-nitrophenyl) carbonate 1 * 1 g
Catalog # PHBLPBY0353-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, EMPLURA®
Supplier: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals