3-[2-[2-[5-[rel-(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethoxy]ethoxy]propanoic acid 1 * 1 g
Catalog # PHBLPBY0278-1G
Supplier: Pharmablock (USA)
3-[2-[2-[5-[rel-(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]ethoxy]ethoxy]propanoic acid 1 * 1 g
Catalog # PHBLPBY0278-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Chloroform, extra dry 99.9% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.8%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, anhydrous ≥99.8%, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) for histology
Supplier: Honeywell Chemicals
Ethanol 80% vol, Molecular Biology Grade
Supplier: VWR Chemicals
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific